missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ E-Gel™ Power Snap Electrophoresis Device Starter Kit, EX 1%

Product Code. 15716558
Change view
Click to view available options
Quantity:
1 Kit
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15716558 1 Kit Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: This product is under eco-contribution tax.
Product Code. 15716558 Supplier Invitrogen™ Supplier No. G8141ST

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product is under eco-contribution tax.

Includes

E-Gel Power Snap Electrophoresis device,orange viewing glasses,E-Gel Go! agarose gel adaptor,region specific cable with power adaptor.

Designed for fast and convenient E-Gel™ agarose gel electrophoresis, from DNA sample loading to gel image. Includes E-Gel EX 1% agarose gels.

The Invitrogen E-Gel Power Snap Electrophoresis Device Starter Kit, EX 1%, provides everything you need to quickly start using precast E-Gel EX 1% agarose gels on the device.

The kit includes:

  • 10 E-Gel EX agarose gels, 1% (Cat. No. G401001)
  • E-Gel Power Snap Electrophoresis Device (Cat. No. G8100)
  • E-Gel 1 Kb Plus Express DNA Ladder with E-Gel Sample Loading Buffer (1X) (Cat. No. 10488091)
  • Gel opening knife (Cat. No. EI9010)

Warranty

1 year warranty

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage • 1 electrophoresis device
• 10 E-Gel EX 1% gels
• 1 vial E-Gel 1 Kb Plus Express DNA Ladder
• 1 power adapter
• 1 pair of viewing glasses
• 1 knife

Store at room temperature.
Type Electrophoresis System Starter Kit
Quantity 1 Kit
Shipping Condition Room Temperature
Max. Voltage 48 VDC
Voltage 240 VAC

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.