missing translation for 'onlineSavingsMsg'
Learn More

Contec™ ProChlor
BIOPHARMA

Product Code. 15904332 Shop All Contec Products
Change view
Click to view available options
Container Type:
Capped Bottle
Trigger Spray Bottle
Quantity:
2 x 5 L
6 x 1 L
Sterility:
Non-sterile
Sterile
Unit Size:
2 bottles
6 L
6 bottles
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Container Type Quantity Sterility unitSize
15904332 Capped Bottle 6 x 1 L Sterile 6 bottles
15537161 Capped Bottle 2 x 5 L Non-sterile 2 bottles
15547171 Trigger Spray Bottle 6 x 1 L Sterile 6 bottles
15567161 Trigger Spray Bottle 6 x 1 L Non-sterile 6 bottles
15597161 Capped Bottle 2 x 5 L Sterile 2 bottles
16479760 Capped Bottle 6 x 1 L Non-sterile 6 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 15904332 Supplier Contec™ Supplier No. SBC102PC

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Contec ProChlor is a revolutionary new sporicide achieving a 100% kill against spores in under 1 min.

Ensures cleanroom is free from contamination and particulates.

A blend of hypochlorous acid in purified water, Contec ProChlor is provided ready-to-use and is efficacious against bacteria, fungi, moulds, yeasts, viruses and spores.

Contec ProChlor is 0.2 micron filtered, and filled in a Grade A environment. Provided double bagged, the product is designed for ease of entry into pharmaceutical cleanrooms.

Features & Benefits

  • Log 6 kill against Bacillus subtilis in 60 secs saves time spent on biodecontamination
  • Filtered to 0.2 microns in a Grade B cleanroom ensures the product is free from contamination and particulates
  • Sterile version available for Grade A and B cleanrooms
  • Contains no quaternary ammonium or surfactant so it's very low residue
  • No hazard classification means only basic PPE required and no special disposal is required
  • Double bagged packed in linear tear packaging so it's easy to open even when wearing gloves
  • Not classed as corrosive and can be used safely in all areas of the cleanroom
Note: For trigger spray option, see 15567161.
TRUSTED_SUSTAINABILITY

Specifications

Product Type Disinfectant
Form Liquid
pH 3 to 6
Sterility Sterile
Appearance Colorless
Autoclavable Not Autoclavable
Certifications/Compliance Grade A and B cleanrooms
Color Clear
Composition Fluid
Container Type Capped Bottle
Disposable Limited-use
Filtered Filtered
Flammable No
Quantity 6 x 1 L
Volume (Metric) 1 L
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.