missing translation for 'onlineSavingsMsg'
Learn More

Miele Professional™ ProCare Lab 10 MA - 5 L

Liquid detergent, mildly alkaline, 5 L. For optimum reprocessing of laboratory glassware and utensils.

Brand:  Miele Professional™ 10265670


Product Code. 15430425

  • 1000.00 DKK / 5 L

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

  • Free from surfactants and oxidants
  • Specially developed for Miele Professional
  • Suitable for all types of materials
  • pH value 11.6 to 11.8
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Liquid Detergent
Liquid detergent, mildly alkaline
5 L
11.6 to 11.8
For optimum reprocessing of laboratory glassware and utensils
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Miele ProCare Lab 10 MA

Signal Word
  • Danger

Hazard Category
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 B

Hazard Statement
  • H290-May be corrosive to metals.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

Supplemental information
  • MIXTURE LIST-Contains : disodium metasilicate

SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.