missing translation for 'onlineSavingsMsg'
Learn More

Primary Opalescent Suspension, EP Grade, Reagecon™
BIOPHARMA

Product Code. 11835853 Shop All Reagecon Products
Change view
Click to view available options
Quantity:
100 mL
Unit Size:
100mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11835853 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11835853 Supplier Reagecon Supplier No. EPPOS01

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Used to calibrate, control, qualify and validate turbidimeters and nephelometers. Primary Opalescent Suspension, EP Grade, Reagecon™ is a ready-to-use formazin suspension with an opalescent value of 4000 NTU. It is designed for use as a turbidity standard in accordance with Eu. Ph. Chapter 2.

This Primary Opalescence Suspension is a ready-to-use turbidity standard presented in high-quality, tamper-evident bottles for use in calibrating turbidimeters and nephelometers and for controlling the reproducibility of their measurements.

This product is a formazin suspension prepared with equal parts hexamethylenetetramine solution and hydrazine sulphate solution. Aqueous formazine solutions feature small, uniform colloidal particles that scatter light in all directions, causing opalescence.

  • Manufactured according to European Pharmocopoeia Chapter 2(2.2.1) guidelines with a Balance Certificate of Calibration issued in accordance with ISO 17025
  • Prepared gravimetrically on a weight/weight basis using OIML traceable weights
  • Concentration verified using a high-performance spectrophotometer calibrated using ISO Guide 34 accredited spectrophotometric standards
  • The Primary Opalescence Suspension diluted in precise ratios to form the EP Reference Opalescence Suspensions (I, II, III, IV)

Specifications

Chemical Name or Material Formazin suspension
Description Opalescence value = 4000 NTU
CAS 100-97-0, 10034-93-2, 7732-18-5
Certificate of Analysis Yes
Analyte or Component Names Hexamethylenetetramine solution + Hydrazine sulphate solution
Physical Form Liquid Suspension
Synonym Formazin suspension
Element Hexamethylenetetramine, Hydrazine Sulphate
Quantity 100 mL
Grade Eu. Ph. Chapter 2
compliance-icons compliance-icons
Product Identifier
  • PRIMARY OPALESCENT SOLUTION
Signal Word
  • Danger
Hazard Category
  • Skin sensitiser Category 1
  • Carcinogenicity Category 1B
  • Skin sensitiser Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H350-May cause cancer.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P321-Specific treatment (see on this label).
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: methenamine, hydrazinium(2+) sulphate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.