Learn More
Proteintech Prestained Protein Marker
The PL00001 Regular Range Protein Marker is a ready to use three-color protein standard with 10 prestained proteins covering a wide range of molecular weights from 10 to 180 kDa.
Brand:  Proteintech PL00001-500UL
Product Code. 17398252
  • € 119.00 / 500µL
Please to purchase this item. Need a web account? Register with us today!
This item is not returnable. View return policy
Description
Description
  • The PL00001 Prestained Protein Marker is designed for monitoring protein separation during SDS-polyacrylamide gel electrophoresis, verification of Western transfer efficiency on membranes, (PVDF, nylon, or nitrocellulose) and for estimating the size of proteins.
  • Contents: Approximately 0.1~0.4 mg/mL of each protein in the buffer (20 mM Tris-phosphate (pH 7.5 at 25°C), 2 % SDS, 0.2 mM Dithiothreitol, 3.6 M Urea, and 15 % (v/v) Glycerol).
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates