Learn More
Sampling Systems™ SteriWare™ Disposable Powder Spatula
BIOPHARMA
Product Code. 12554366 Shop All Sampling Systems Products
Change view
Click to view available options
Length (Metric):
230 mm
Unit Size:
Pack of 100
Product Code. Length (Metric) unitSize
12554366 230 mm Pack of 100
1 options
Product Code. 12554366 Supplier Sampling Systems™ Supplier No. 8095A150S
Only null left
Add to basket
Edge
Add to basket
Tough and comfortable to use with its specially designed rounded handle and robust blade. The Powder Spatula is manufactured and packed in a controlled atmosphere (class 100,000 cleanroom) and so can be used straight from the bag.
Key Features:
  • Moulded and packed in a Class 100,000 Cleanroom
  • Individually bagged
  • Made from FDA, EC 1935/2004 and EU 10/2011 conforming material
  • BSE/TSE free
  • Full batch traceability
  • Available pre-sterilized (SAL 10-6)
  • Suitable for use in ATEX Zones 0, 1 and 2
  • Available in 2 different sizes and a variety of colors
Why Use a SteriWare™ Disposable Sampler?
  • No Cross Contamination - All SteriWare™ devices can be used straight from the bag.
  • No Cleaning - Unlike a reusable device, you can use a SteriWare™ product straight away. This eliminates the need for costly Cleaning Validation.
  • High Quality - Our SteriWare™ products are manufactured/moulded/packed in a Class 100,000 Cleanroom in England. Please see the products Certificate of Conformity and Technical Specification Sheet for more information.
  • Full Documentation - We can provide full batch specific documentation for all SteriWare™ products. This includes Certificates of Conformity and Certificates of Irradiation (where applicable). The batch number is clearly printed on every bag and box.
  • Cost Effective - Removing the need for Cleaning Validation by switching to a SteriWare™ device saves time and money.
  • Available Sterile - Most SteriWare™ products are available pre-sterilized by Gamma or Electron Beam to SAL10-6. Please see the Specification Sheet for more information.
Order Info
LengthMetric
Specifications
Quantity 100 Pk.
Sterility Sterile
Color White
Disposable Single-use
Product Type Powder Spatula
Material Polystyrene
Length (Metric) 230 mm