missing translation for 'onlineSavingsMsg'
Learn More

Polysorbate 80, Multi-Compendial, N.F., J.T. Baker™

Product Code. 15469933 Shop All J.T.Baker Products
Change view
Click to view available options
Quantity:
20 mL
200 mL
1 L
4 L
19 L
Packaging:
Narrow Mouth Amber Glass Bottle
Narrow Mouth Amber Glass Bottle, w/ Septum
Steel Drum
Unit Size:
1 L
19 L
200mL
20mL
4 L
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15469933 20 mL Narrow Mouth Amber Glass Bottle 20mL
10554683 200 mL Narrow Mouth Amber Glass Bottle 200mL
15528894 1 L Narrow Mouth Amber Glass Bottle 1 L
12615457 1 L Narrow Mouth Amber Glass Bottle, w/ Septum 1 L
15362724 4 L Narrow Mouth Amber Glass Bottle 4 L
15538894 19 L Steel Drum 19 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 15469933 Supplier Avantor J.T.Baker Supplier No. 411702

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 9005-65-6
Molecular Formula (C2H4O)x(C2H4O)z(C2H4O)y(C2H4O)wC24H44O6
Molecular Weight (g/mol) 604.82
MDL Number MFCD00082107
InChI Key HDTIFOGXOGLRCB-KTKRTIGZNA-N
IUPAC Name 2-{2-[3,5-bis(2-hydroxyethoxy)oxolan-2-yl]-2-(2-hydroxyethoxy)ethoxy}ethyl (9E)-octadec-9-enoate
SMILES CCCCCCCC\C=C/CCCCCCCC(=O)OCCOCC(OCCO)C1OCC(OCCO)C1OCCO

Specifications

Quantity 20 mL
Flash Point 148.00°C
Packaging Narrow Mouth Amber Glass Bottle
Specific Gravity 1L = 1.08kg
Grade Multi-Compendial/NF
Chemical Name or Material Polysorbate 80
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.