missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
- Manufactured from polypropylene
- Includes tip tray
- Empty, for filter and pipet tips
Specifications
Specifications
| Color | Yellow |
| Holds | Pipette Tips (200 μL) |
| Material | Polypropylene |
| Stackable | Yes |
| For Use With (Equipment) | Pipet and Filter Tips |
| No. per Pack | 1 Pack |
| Quantity | 1 Pack |
| Product Type | Empty Tip Box |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?
