missing translation for 'onlineSavingsMsg'
Learn More

Eppendorf™ Polypropylene Protein LoBind Microcentrifuge Tube
GREENER_CHOICE

Code produit 10708704 Tous les produits Eppendorf Produits
Change view
Click to view available options
Capacity (Metric):
0.5 mL
1.5 mL
2 mL
Conditionnement:
Pack of 100
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Capacity (Metric) unitSize
10708704 1.5 mL Pack of 100
10316752 0.5 mL Pack of 100
10718894 2 mL Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10708704 Supplier Eppendorf™ Supplier No. 0030108116
Only null left
Add to basket
Edge
Add to basket
This item is not returnable. View return policy

Store peptide, antibody or virus samples in these LoBind microcentrifuge tubes. Eppendorf™ Polypropylene Protein LoBind Microcentrifuge Tubes are ideal for work in protein research, mass spectrometry and protein array sample preparation.

  • Problem free handling with the tiniest volumes/concentrations of a variety of proteins
  • Preliminary handling of tubes by saturation with BSA or siliconisation is not required in order to guarantee the recovery of the proteins in the plastic tube
  • Recovery rates of more than 99% are achievable
  • Enzyme activities, for example, are not reduced by this tube, and denaturing effects caused by interaction with the surface are almost completely excluded
  • No undesired side effects of a surface coating
  • Tubes are available in Eppendorf LoBind™ material for maximum recovery of valuable samples
  • Maximum protein recovery (over 97%, BSA 1μg/mL)
  • Free from surface coating, e.g., silicone
  • Free from DNA, DNase, RNase and ATP (PCR* clean)
  • Special polypropylene material manufactured under optimized processing conditions
  • High degree of centrifugation durability, rated to 18, 000xg
  • High degree of transparency
  • Safe-Lock lid
TRUSTED_SUSTAINABILITY

Specifications

Closure Material Polypropylene
Capacity (Metric) 1.5 mL
Color Transparent
Material Polypropylene
For Use With (Application) Protein LoBind consumables are best suited for work in protein research, mass spectrometry, protein array sample preparation, and storage of peptide, antibody or virus samples
No. per Pack 100 Tubes (2 bags of 50)
Volume (Metric) 1.5mL
Sterility PCR Clean
Max. RCF 18,000 x g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.