missing translation for 'onlineSavingsMsg'
Learn More

Kimberly-Clark™ Kimtech™ Process Wipers 7622, 35 Quarter-folded

Designed to be strong, low-linting and 100% silicon-free. Great for wiping oils and grease, and repeat-use cleaning with chemical and solvents. Cloths are folded twice to provide maximum efficiency. Packaged in a splash-proof, water repellent polybag. Quantity 12 packs x 35 quarter-folded sheets, blue (420 total).

Brand:  Kimberly-Clark™ 7622

2050.38 DKK valid until 2025-12-31
Use promo code "25339" to get your promotional price.


Product Code. 10728571

  • 3325.00 DKK / Pack of 12
Only null left
In Stock
Edge
Add to basket

Description

Description

  • 12 x 35 quarter-folded, blue sheets (420 total)
  • 100% silicon free and low-linting
  • Great for wiping oils and grease, and repeat-use cleaning with chemical and solvents
  • Cloths are folded twice to provide maximum efficiency
  • Packaged in a splash-proof, water repellent polybag
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Process Wiper
52.3 cm
40.2 cm
Blue
Quarter Fold
1-ply
15.74 in.
15.74 in
Polypropylene
380 x 490 mm
420 Pk.
Non-sterile
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.