missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Grant Instruments™ Polyethylene Draining Syphon
For Grant heated circulating baths
Brand: Grant Instruments™ SY1
This item is not returnable.
View return policy
Description
- For use with Grant heated circulating baths
Specifications
| Discharge Rate: 6.5L/min. | |
| Discharge Hose Size: 21.7in. | |
| Container-side Hose Size: 15 in. |
| Drain liquids from Grant water bath for 5 to 20L (5 gal.) containers | |
| Polyethylene |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction