missing translation for 'onlineSavingsMsg'
Learn More

Berkshire International™ BCR™ Polyester Half-Finger Cleanroom Glove Liners
BIOPHARMA

Product Code. 12405180 Shop All Berkshire Products
Change view
Click to view available options
:
Medium
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
12405180 Medium
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12405180 Supplier Berkshire International™ Supplier No. BGL2.20R
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Continuous filament knitted polyester construction is thin yet durable, while the half-finger design permits tactility in addition to wicking moisture from the palms.

  • Reduces perspiration and contact dermatitis commonly associated with wearing latex, urethane, PVC, or nitrile gloves
  • Half-finger design permits tactility while also wicking moisture from the palms
  • 100% continuous filament knitted polyester is thin, yet durable
  • Ambidextrous design can be used on either hand
  • Gloves can be repeatedly laundered for reuse
  • Suitable for use in ISO Class Cleanrooms

Specifications

Construction Seamless Knit
Coating Material Uncoated
Color White
Disposable Reusable
Hand Ambidextrous
Material Polyester
Product Type Glove Liners
Size Regular
Style Reversible
Finger Style Half
For Use With (Application) Laboratories and Manufacturing Environments
Glove Cuff Style Knit
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.