Learn More
Berkshire™ Durx™ 670 General Purpose Wipes
BIOPHARMA
Product Code. 12877143 Shop All Berkshire Products
Change view
Click to view available options
Length (Metric):
230 mm
Unit Size:
Pack of 300
Product Code. Length (Metric) unitSize
12877143 230 mm Pack of 300
1 options
Product Code. 12877143 Supplier Berkshire™ Supplier No. DR670.0909.20
Only null left
Add to basket
Edge
Add to basket
Use these wipes for cleaning and maintenance in laboratories as well as aerospace and defense, microelectronics, food processing, pharmaceutical and biomedical device manufacturing environments. Berkshire™ Durx™ 670 General Purpose Wipes feature exceptional strength and absorbency.
  • Manufactured without the use of binders or surfactants, ion levels and organic extractables remain low
  • The same wipe material can be provided in pre-wetted formats for reduced VOC emissions, increased convenience, increased productivity, improved solvent control and cleaning protocol repeatability and reduced costs
  • For aseptic processing areas, the same wipe material can be provided in a gamma irradiated format, validated sterile to a 10–6 sterility assurance level
  • Dry and pre-wetted versions available
  • FDA 21 CFR – Food contact compliant
Specifications
Product Type Cleanroom Wiper
Length (Metric) 230 mm
Width (Metric) 230 mm
Material Polyester Cellulose
Edges Knife-Cut
Packaging Quantity 300 Pk.