Learn More
MP Biomedicals™ Pluronic F-68 Polyol, 10% Solution (100X)
Non-ionic surfactant in cell culture used to control shear forces in suspension cultures against possible damage.
Brand: MP Biomedicals™ 092750049
233.70 DKK valid until 2025-10-31
Use promo code "24120" to get your promotional price.
Description
Pluronic F-68 is a difunctional block copolymer surfactant terminating in primary hydroxyl groups. It is a non-ionic detergent that protects cells from hydrodynamic damage, including freezing and thawing, transfer and stirring. It can also be used to reduce foaming in stirred cultures and reduce cell attachment to glass. Used for in vitro isolation of lymphocytes from diluted whole blood. Pluronic F-68™ is a registered trademark of BASF Corporation.
Specifications
Pluronic F-68™ Polyol | |
100 mL | |
10% Solution, 100X |
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.