Learn More
Macherey-Nagel™ pH-Fix Test Strips in PT Tubes
Product Code. 10025504 Shop All Macherey-Nagel Products
Change view
Click to view available options
pH Range:
0 to 14
Unit Size:
Pack of 100
Product Code. pH Range unitSize
10025504 0 to 14 Pack of 100
1 options
This item is not returnable. View return policy
Product Code. 10025504 Supplier Macherey-Nagel™ Supplier No. 92111
Please to purchase this item. Need a web account? Register with us today!
Edge
This item requires a price update prior to being ordered.
Request a Price for this item here.
This item is not returnable. View return policy
Provides quick and easy pH value testing at the point of interest. Macherey Nagel™ pH-Fix Test Strips in PT Tubes don't need any calibration, set-up or maintenance.
Provides quick and easy pH value testing at the point of interest. Macherey Nagel™ pH-Fix Test Strips in PT Tubes don't need any calibration, set-up or maintenance.
  • Long plastic handle protects against toxic, dangerous or aggressive substances
  • Features up to 4 different indicator pads that cover the whole pH range
  • No bleeding due to color bounded indicator dyes
  • Tube features simple one-hand operation, prevents against jamming, and is virtually unbreakable
Specifications
pH Range 0 to 14
Quantity 100 Pack
Type pH Test Strip