missing translation for 'onlineSavingsMsg'
Learn More

pH Electrode

Product Code. 13121754
Change view
Click to view available options
pH Range:
0 to 14
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. pH Range unitSize
13121754 0 to 14 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 13121754 Supplier Jenway Supplier No. 9996034
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Use these electrodes for advanced applications where faster response times and greater reproducibility are required.

  • Quickly reach equilibrium
  • Significantly reduced response times
  • Far greater reproducibility
  • Significantly reduced long-term drift

Specifications

Dimensions (L x W) 120 x 12 mm
Type pH Combination Electrode
For Use With (Application) General purpose, epoxy bodied for standard measurements of general solutions (max.temperature 60ºC)
Material Epoxy
pH Range 0 to 14
Temperature (Metric) Range 0°C to 60°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.