Learn More
Grant Instruments™ Optima™ Refrigerated Circulating Bath Kit

Provides outstanding performance for routine and sophisticated applications requiring accurate temperature control in the range of -30 to 100°C. Grant Instruments™ Optima™ Refrigerated Circulating Bath Kit includes the TX150 heating circulator and an R4 tank/refrigeration unit, complete with insulated tubing and clips to form a system ready to use.
Brand: Grant Instruments™ LTC4R
7886.03 EUR valid until 2025-12-31
Use promo code "24306" to get your promotional price.
Description
- Powerful pump for external circulation, maximum flow 18 L/min, maximum pressure 310 mbar
- Full colour QVGA TFT display
- Setpoint, plus three adjustable preset temperature values
- Timer function for reaction timing, 99 hours, 59 minutes, with audible alert and heater control
- Liquid selection safety feature adjusts for temperature range or heater power
- Fault indication display
- Programmable relay
- Two-point user recalibration of internal and external probes (TXPEP, TXSEP)
- Off-set adjustment for optimal isothermal performance
- Can be programmed to switch the refrigeration on and off during a heating and cooling program
- Memory capacity for one program of up to 30 segments
- Choice of languages: English, French, German, Spanish and Italian
- Low liquid protection by float switch
- Adjustable over-temperature cut-out
- Convenient accessory clamp available for fitting to vessels
- Stability ± 0.1°C
Specifications
| Kit includes TX150 heating circulator and R4 tank/refrigeration unit | |
| 20 L | |
| -30°C to +100°C | |
| 120/230 V |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.