missing translation for 'onlineSavingsMsg'
Learn More

Sartorius™ Optifit Tips in FlexiBulk Packs
BIOPHARMAGREENER_CHOICE

Product Code. 15310645 Shop All Sartorius Products
Change view
Click to view available options
Volume (Metric):
0.5 to 200 μL
10 to 1000 μL
5 to 350 μL
50 to 1200 μL
Color Code:
Dark Blue
Light Blue
Orange
Purple
Yellow
Length (Metric):
5.1cm
5.4cm
6.85cm
7.15cm
Unit Size:
Pack of 480
Pack of 960
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Color Code Length (Metric) unitSize
15310645 10 to 1000 μL Dark Blue 7.15cm Pack of 480
15300645 5 to 350 μL Orange 5.4cm Pack of 960
15340645 50 to 1200 μL Purple 7.15cm Pack of 480
15390635 0.5 to 200 μL Yellow 5.1cm Pack of 960
15330645 10 to 1000 μL Light Blue 6.85cm Pack of 480
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 15310645 Supplier Sartorius™ Supplier No. LHB791004
Only null left
Add to basket
Edge
Add to basket

Combine cost savings with saving space, improving usability and ensuring purity. Sartorius™ Optifit Tips in FlexiBulk Packs deliver the same high-quality Optifit Tips for use with Biohit™ pipettes in a convenient packaging design. Available in 200, 350, 1000 and 1200μL tip volumes, including a Wide Bore option.

Optifit Tips
  • Perfect fit and seal to ensure the highest possible accuracy and precision
  • Ergonomic and light tip attachment/ejection via the Optiload feature in Sartorius pipettes
  • Color coded for easy matching with suitable Sartorius pipettes
  • Wide bore tips available for better dispensing of difficult samples, such as viscous liquids
  • Compatible with Picus™ NxT, Picus™, eLINE™, mLINE™ and Proline™ Plus pipettes and most other pipette brands
  • Compliant with ISO 9001, ISO 14001 and ISO 13485 quality standards
FlexiBulk Packaging
  • Convenience and savings
    • Combines the low cost of bulk tips with saving space and improving usability
    • Reduces size of packaging up to 40% and cuts packaging waste by nearly 50%
    • Materials in the FlexiBulk pack are 100% suitable to be used as energy waste
    • Pick & Place organization for quick and easy rack loading direct tip attachment
    • Easy open/close lid for easy tip access and for protecting tips
  • Quality and purity
    • Includes the exact same, high-quality tips as other packaging options
    • Ensures tip purity with lot-specific certificates
    • Nonsterile tips are certified free of RNase, DNase and endotoxins
    • Manufactured under ISO Class 8 cleanroom conditions
    • Lid provides an airtight seal during transport and storage for secured purity
TRUSTED_SUSTAINABILITY

Specifications

Volume (Metric) 10 to 1000 μL
Length (Metric) 7.15cm
Certifications/Compliance ISO 9001, ISO 14001, ISO 13485
Clean Claims DNase, RNase, ATP, and Endotoxin Free
For Use With (Application) For everyday laboratory work
Format Bulk
Color Code Dark Blue
For Use With (Equipment) Sartorius Picus NxT, Sartorius Picus, Sartorius eLINE, Sartorius mLINE, Sartorius Proline Plus, Sartorius Biohit
Product Line Optifit FlexiBulk
Material Polypropylene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.