missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Matrix™ Open-Top Tri-coded Tubes
GREENER_CHOICE

Securely store and track samples in Thermo Scientific™ Matrix™ Open-Top Tri-coded tubes. Tubes feature permanent, high contrast 2D barcodes on the bottom, plus 1D linear barcodes with human readable text on the side.

6310.00 DKK - 8025.00 DKK

Specifications

Product Type Storage Tube (Open Top)
Autoclavable Not Autoclavable
Product Line Matrix™
Graduated No
View More Specs

Products 4
Product Code Brand Capacity (Metric) Sterility Packaging Price Quantity & Availability  
Product Code Brand Capacity (Metric) Sterility Packaging Price Quantity & Availability  
17278820
GREENER_CHOICE
View Documents Promotion Details
Thermo Scientific™
3790-WP1D
1400 μL Non-sterile Bulk
7015.00 DKK
Pack of 1000
Only null left
Estimated Shipment: 21-10-2025
Add to basket
 
17288820
GREENER_CHOICE
View Documents Promotion Details
Thermo Scientific™
3791-WP1D-BR
1400 μL Non-sterile 10 Latch Racks of 96 Tubes/Cs.
8025.00 DKK
Pack of 480
Only null left
Estimated Shipment: 25-11-2025
Add to basket
 
17298820
GREENER_CHOICE
View Documents Promotion Details
Thermo Scientific™
3792-WP1D-BR
1400 μL Sterile 10 Latch Racks of 96 Tubes/Cs.
8005.00 DKK
Pack of 480
Only null left
Estimated Shipment: 08-08-2025
Add to basket
 
18877980
GREENER_CHOICE
View Documents
Thermo Scientific™
3732-WP1D
750 μL Sterile 10 Latch Racks of 96 Tubes/Cs.
6310.00 DKK
Pack of 10
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Securely store and track samples in Thermo Scientific™ Matrix™ Open-Top Tri-coded tubes. Tubes feature permanent, high contrast 2D barcodes on the bottom, plus 1D linear barcodes with human readable text on the side. Tubes are available in bulk packs or in barcoded latch racks. Barcoded latch racks feature a linear barcode, 2D barcode, and human readable rack identification on three sides of the rack. Use in combination with Thermo Scientific™ Matrix™ SepraSeal and DurSeal capping systems and the Thermo Scientific™ VisionMate™family of barcode readers.

Secure Tracking

  • Each polypropylene open-top tube has a unique 2D barcode laser-etched onto the base, plus a white patch with 1D linear barcode and human readable text on the side
  • 2D barcodes are resistant to 100% DMSO, ideal for compound processing and storage
  • Non-proprietary 2D barcodes and 1D linear barcodes can be scanned with the Thermo Scientific VisionMate family of barcode readers

Superior Storage Format

  • ANSI-format latch racks maximize storage space and are compatible with multichannel manual pipettes or automated liquid handling equipment
  • Barcoded latch racks include 1D and 2D barcodes, plus human readable text on three sides to provide added same traceability
  • The latch rack lid can be lifted robotically for 2D tube access with automated liquid handling systems and high throughput applications

Stringent Quality Control

  • Every 2D barcoded storage tube is scanned to guarantee readability
  • Each code is checked against a full database of all previously assigned 2D codes to ensure zero duplicates across the entire line of Matrix 2D barcoded storage tubes

Storage Tracking Solutions for Any Laboratory

  • Custom 2D coding options are available to meet your laboratory's individual database or tracking requirements
  • Tubes are available with a range of sealing options including Thermo Scientific SepraSeals and DuraSeals
  • SepraSeals are available in a range of color options for visual categorization of samples
  • Pre-Split SepraSeals allow for repeated sample access in automation

Recommended for:

Drug discovery, compound storage, secure sample transport, individual sample retrieval, instant sample identification, secure tracking of archived samples

Specifications

Specifications

Storage Tube (Open Top)
Not Autoclavable
Matrix™
No
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.