missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Oakton™ Testr PCTS 50 Waterproof Pocket pH/Cond/TDS/Salinity Tester
Great for water, wastewater, laboratory or plant use, these multiparameter testers feature long-life pH electrodes with a wide range of sample compatibility. Ideal for pH, conductivity, TDS, salinity, and temperature testing.
Merk: Oakton™ 35634-35
Includes: Protective plastic storage case, lanyard, and batteries.
Beschrijving
- Combine measurement capabilities of three testers in one convenient pocket-sized unit
- Up to 3 point pH calibration with NIST or USA auto buffer recognition
- Conductivity, TDS, and salinity feature auto ranging based on the concentration of your sample
- Redesigned leakproof cap functions as a sample/calibration cup, hydrating cup, and stabilizing base to stand the meter upright
Services
1 Year
Specificatie
±1% full scale | |
±1% full scale | |
150+ hours | |
Up to 3 points | |
1 | |
IP67 | |
0 to 20.00 mS | |
0 to 10.00 ppt | |
0.01 | |
Water quality testing, buffer and reagent production, electrochemistry | |
Membrane | |
Hold Data | |
Integral multiparameter probe | |
0.1 μS, 1 μS; 0.01 mS | |
Checkmark indicator | |
Automatic | |
5°C to 45°C | |
0.1°F/°C |
±0.01 | |
Yes | |
4 AAA Alkaline | |
3 | |
Graphics, Dot Matrix 80 x 100 pixel | |
pH, Conductivity, TDS, Salinity, Temperature | |
-1 to 15 | |
0 to 10.00 ppt | |
0°C to 60°C | |
Protective plastic storage case, lanyard, and 4 AAA batteries. | |
English | |
50 Series | |
Salinity: 0.10 ppt | |
0.1 ppm, 1 ppm, 0.01 ppt | |
±0.9°F (0.5°C) from 32 to 122°F (0 to 50°C); ±1.8°F (1.0°C) from 122 to 140°F (50 to 60°C) | |
41°F to 113°F (ambient) | |
32°F to 140°F | |
1 Year |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction