missing translation for 'onlineSavingsMsg'
Learn More

o-Xylene-d{10}, 98+% (Isotopic)

Product Code. 15401457
Click to view available options
Quantity:
5 g
Unit Size:
5g
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

CAS: 56004-61-6 | C8H10 | 116.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Physical Form Liquid
Color Colorless
Density 0.953 g/mL
Sensitivity Moisture
Quantity 5 g
CAS 56004-61-6
Melting Point -25°C
Boiling Point 143°C to 145°C
Molecular Formula C8H10
MDL Number MFCD00037698
UN Number UN1307
Merck Index 14,10081
Synonym o-xylene-d10, 2h10-o-xylene, 1,2-dimethylbenzene-d10 99.5 atom % d, 1,2-xylene-d10, o-xylene-d10, 99 atom % d, bis 2 h? methyl 2 h? benzene, 1,2,3,4-tetradeuterio-5,6-bis trideuteriomethyl benzene
InChI Key CTQNGGLPUBDAKN-ZGYYUIRESA-N
SMILES [2H]C1=C([2H])C([2H])=C(C(=C1[2H])C([2H])([2H])[2H])C([2H])([2H])[2H]
IUPAC Name 1,2,3,4-tetradeuterio-5,6-bis(trideuteriomethyl)benzene
Molecular Weight (g/mol) 116.23
PubChem CID 91976
Formula Weight 116.25
Percent Purity ≥98%
Chemical Name or Material o-Xylene-d{10}, Isotopic
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.