Learn More
Beschreibung
- NuGenius XE offers flexible DNA, stain free, autorads analysis.
- Intuitive touch screen system.
- Compact benchtop size with built in PC.
- Internal lighting options includes a UV transilluminator (purchased separately) for working with DNA gels (Ethidium bromide).
- Our new UV-blue light converter screen (purchased separately) allows imaging of a wide range of safe dyes.
- A visible converter option (purchased separately) can quickly extend its use for working with visible gels and blots (Coomassie blue and Silver stain).
- The overhead LED white lighting is included as standard for easy sample positioning and focussing.
- Safely excise your DNA bands from an agarose gel with the darkroom door open using the optional safety shield protecting the user when working with UV.
- With a 5M pixel camera and f/1.2 motor driven lens provides exquisite pixel resolution and unrivalled sensitivity for gels and blots with a viewing area of 20 x 24 cm.
- Images are saved directly to USB or network for easy transfer.
- Quickly generate quantitative data with GeneTools analysis software.
- NuGenius XE comes with unlimited copies of GeneTools.
Spezifikation
Spezifikation
| Display | Touchscreen |
| Pixelgröße | 5 MP |
| Zur Verwendung mit (Anwendung) | Gel Documentation |
| Produkttyp | Gel-Bildgebungssystem |
Name des Produkts