missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ NucleoSpin™ RNA Virus 50 Preps

Product Code. 12791021
This item is not returnable. View return policy
Product Code. 12791021 Supplier Macherey-Nagel™ Supplier No. 740956.50

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mini spin kit for the isolation of viral RNA and DNA from cell-free biological fluids

NucleoSpin™ RNA Virus is designed for the isolation of viral nucleic acids from serum, plasma or any cell-free biological fluids, featuring a special type of membrane which has a high binding capacity for nucleic acids.
The procedure is rapid and easy. The lysate is incubated for 5 to 10min and applied to the NucleoSpin™ column. After two washing steps to remove contaminations the RNA can be eluted with water or low-salt buffer. The kit contains all necessary buffers, spin columns and detailed protocols. Generally, NucleoSpin™ RNA Virus is suited for the parallel purification of viral RNA and DNA (support protocol). For example, isolation of HBV DNA is performed with an additional proteinase K digest (enzyme not included in the kit). For isolation of viral DNA with highest sensitivities, the NucleoSpin™ Blood kit is recommended.

Kit components: NucleoSpin™ RNA Virus columns, 2mL collecting tubes, buffers, RNase-free water, carrier RNA.
• Silica membrane technology
• Typical recovery: >90%
• Sample material: 150μL serum, plasma, cell-free biological fluids
• Fragment size: 100bp to 30kb
• Elution volume: 50μL
• Binding capacity: 40μg
• Preparation time: 30min/4-6 preps
• Format: mini spin column
• Support protocol for parallel isolation of viral RNA and DNA (proteinase K required)
• Applicable for HCV, HBV, HAV, HIV, HSV, HPV, VZV, EBV, parvovirus B19, H5N1

Specifications

Content And Storage 18°C to 25°C
Format Mini Prep
Sample Type Cell-free Biological Fluids, Plasma, Serum
For Use With (Application) Isolation of Viral Nucleic Acids
No. of Reactions 50 Preps
Size 100 bp−approx. 50 kbp
Target Viral RNA and DNA
Volume (Metric) < 150 L
compliance-icons compliance-icons
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P264-Wash with water thoroughly after handling.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P330-Rinse mouth.
Supplemental information
  • MIXTURE LIST-Contain: guanidine hydrochloride, ethanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.