missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Macherey-Nagel™ NucleoSpin™ Gel and PCR Clean-up, kit for gel extraction or PCR clean up
Click to view available options
No. of Reactions:
240 Preps
50 Preps
Unit Size:
240 preps
50 reactions
Description
NucleoSpin™ Gel and PCR Clean-up is designed for fast purification of PCR products, such as DNA from enzymatic reactions, as well as the extraction of DNA fragments from TAE or TBE agarose gels.
- Two applications in one kit—One buffer to process both sample types, with only one loading step
- High recovery for fragments down to 50 bp—Average recovery from 60–90%
- Minimized elution volume of 15μL—Highly concentrated DNA
- Binding buffer with pH indicator—Ensures optimal binding conditions
- Separate buffers for single stranded DNA and RNA or for SDS-containing samples available separately
- Suitable for all kinds of agarose gels and a variety of buffer systems (e.g. TAE, TBE)
- Complete removal of contaminations: nucleotides, primers, enzymes, mineral oil, PCR additives, detergents, dyes
- Applications: purification of PCR products, extraction of DNA from agarose or polyacrylamide gels, and downstream applications (cloning, sequencing, PCR, restriction analysis)
Kit contents: NucleoSpin™ Gel and PCR Clean-up columns, collection tubes (2mL), binding buffer with pH indicator, wash buffer and elution buffer
Specifications
Specifications
Color | pH Indicator: Yellow = correct conditions Green = pH slightly high (adjustment recommended) Blue = pH too high (adjustment necessary) |
Format | Mini Spin Column |
Isolation Technology | Silica Membrane Technology |
Efficiency | Typical recovery: 70–95% |
Elution Volume | Elution volume: 30–100μL |
For Use With (Equipment) | QIAcube™ compatible TAE or TBE agarose gels, PCR |
Includes | Binding Buffer NTI (2 x 75mL), Wash Buffer NT3 Concentrate (60mL), Elution Buffer NE (125mL), NucleoSpin Gel and PCR Clean-up Columns (4 x 60) |
No. of Reactions | 240 Preps |
Quantity | 1 Kit |
Purity | A260/A280: 1.8–1.9 |
Show More |
Safety and Handling

Signal Word
- Warning
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 3
- H302-Harmful if swallowed.
- H412-Harmful to aquatic life with long lasting effects.
- MIXTURE LIST-Contains : guanidinium thiocyanate
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction