Learn More
Macherey-Nagel™ NucleoSpin™ Gel and PCR Clean-up, kit for gel extraction or PCR clean up
Description
NucleoSpin™ Gel and PCR Clean-up is designed for fast purification of PCR products, such as DNA from enzymatic reactions, as well as the extraction of DNA fragments from TAE or TBE agarose gels.
- Two applications in one kit—One buffer to process both sample types, with only one loading step
- High recovery for fragments down to 50 bp—Average recovery from 60–90%
- Minimized elution volume of 15μL—Highly concentrated DNA
- Binding buffer with pH indicator—Ensures optimal binding conditions
- Separate buffers for single stranded DNA and RNA or for SDS-containing samples available separately
- Suitable for all kinds of agarose gels and a variety of buffer systems (e.g. TAE, TBE)
- Complete removal of contaminations: nucleotides, primers, enzymes, mineral oil, PCR additives, detergents, dyes
- Applications: purification of PCR products, extraction of DNA from agarose or polyacrylamide gels, and downstream applications (cloning, sequencing, PCR, restriction analysis)
Kit contents: NucleoSpin™ Gel and PCR Clean-up columns, collection tubes (2mL), binding buffer with pH indicator, wash buffer and elution buffer
Specifications
Specifications
| Color | pH Indicator: Yellow = correct conditions Green = pH slightly high (adjustment recommended) Blue = pH too high (adjustment necessary) |
| Format | Mini Spin Column |
| Isolation Technology | Silica Membrane Technology |
| Efficiency | Typical recovery: 70–95% |
| Elution Volume | Elution volume: 15–30μL |
| For Use With (Equipment) | TAE or TBE agarose gels, PCR |
| Includes | Binding Buffer NTI (40mL), Wash Buffer NT3 Concentrate (25mL), Elution Buffer NE (13mL), NucleoSpin Gel and PCR Clean-up Columns (50), 2mL Collection Tubes (50) |
| No. of Reactions | 50 Preps |
| Quantity | 1 Kit |
| Purity | A260/A280: 1.8–1.9 |
| Show More |
Safety and Handling
- Warning
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 3
- H302-Harmful if swallowed.
- H412-Harmful to aquatic life with long lasting effects.
- MIXTURE LIST-Contain: guanidinium thiocyanate
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.