missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Macherey-Nagel™ NucleoSpin™ Gel and PCR Clean-up Columns for gel extraction and PCR clean up
Description
- Columns only (no buffers) for PCR purification or gel extraction applications.
- In accordance with Macherey-Nagel's Quality Management System, each lot of product is tested against predetermined specifications to ensure consistent product quality.
Specifications
Specifications
| Format | Mini Spin Column |
| Isolation Technology | Silica Membrane Technology |
| Efficiency | Typical recovery: 70–95% |
| Elution Volume | Elution volume: 15–30μL |
| For Use With (Equipment) | For use with NucleoSpin kits. |
| Includes | NucleoSpin Gel and PCR Clean-up Columns (250) |
| Quantity | 250 Columns |
| Purity | A260/A280: 1.8–1.9 |
| Sample Volume | <400 μL PCR reaction mixture, <400 μg agarose gel |
| Size | 50 bp to ∼20 kbp |
| Show More |
Safety and Handling
Supplemental information
- MIXTURE LIST-Contain: guanidinium thiocyanate
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?