missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ NucleoSpin™ Gel and PCR Clean-up Kit

Clean up PCR reactions and gel extractions with a two-in-one approach. Macherey-Nagel™ NucleoSpin™ Gel and PCR Clean-up Kit is optimized for recovery and elution volume when purifying DNA fragments via both applications.

Marca:  Macherey-Nagel™ 740609.250


Código de producto. 11992242

  • ONLINE EXCLUSIVE PRICE
    667.00 €
    535.50 € / 250 reactions
    Save 131.50 €
    19% Off

Please to purchase this item. Need a web account? Register with us today!

Edge
Este artículo no se puede devolver. Vea la política de devoluciones

Descripción

Descripción

NucleoSpin™ Gel and PCR Clean-up is designed for fast purification of PCR products, such as DNA from enzymatic reactions, as well as the extraction of DNA fragments from TAE or TBE agarose gels.

  • Two applications in one kit—One buffer to process both sample types, with only one loading step
  • High recovery for fragments down to 50 bp—Average recovery from 60–90%
  • Minimized elution volume of 15μL—Highly concentrated DNA
  • Binding buffer with pH indicator—Ensures optimal binding conditions
  • Separate buffers for single stranded DNA and RNA or for SDS-containing samples available separately
  • Suitable for all kinds of agarose gels and a variety of buffer systems (e.g. TAE, TBE)
  • Complete removal of contaminations: nucleotides, primers, enzymes, mineral oil, PCR additives, detergents, dyes
  • Applications: purification of PCR products, extraction of DNA from agarose or polyacrylamide gels, and downstream applications (cloning, sequencing, PCR, restriction analysis)

Kit contents: NucleoSpin™ Gel and PCR Clean-up columns, collection tubes (2mL), binding buffer with pH indicator, wash buffer and elution buffer

Sugerencias de productos

Sugerencias de productos

Videos
Seguridad y manipulación

Seguridad y manipulación

Palabra de advertencia
  • Warning

Clasificación
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 3

Indicaciones de peligro
  • H302-Harmful if swallowed.
  • H412-Harmful to aquatic life with long lasting effects.

Otros peligros
  • MIXTURE LIST-Contain: guanidinium thiocyanate

SDS
Documentos

Documentos

Product Certifications
Promociones

Promociones

Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.