missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ELISA Assay Buffer (5X)

Product Code. 10518383
Change view
Click to view available options
Quantity:
200 mL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10518383 200 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy

For Research Use Only. All usage must comply with product instructions.

Product Code. 10518383 Supplier Invitrogen™ Supplier No. DS98200

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

For Research Use Only. All usage must comply with product instructions.

For use with antibody pairs (ELISA)

Concentrated 5X Assay Buffer for Antibody Pairs is intended to be used as:

  • Blocking reagent for ELISA plates
  • Diluent for ELISA standards and samples
  • Diluent for the detection antibody
  • Diluent for the HRP conjugate

Build Your Own Immunoassay, Cell Analysis, Immunoassays

Order Info

Shipping Condition: Wet Ice

TRUSTED_SUSTAINABILITY

Specifications

Applications ELISA
Quantity 200 mL
Content And Storage Buffer. Store at 2-8°C.
Form Concentrated
Shipping Condition Wet Ice
compliance-icons
Product Identifier
  • ELISA Assay Buffer (5X)
Signal Word
  • Warning
Hazard Category
  • Skin sensitiser Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P272-Contaminated work clothing should not be allowed out of the workplace.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
  • P362+P364-Take off contaminated clothing and wash it before reuse.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: 5-chloro-2-methyl-3(2H)-isothiazolone, 2-methyl-3(2H)-isothiazolone

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.