Learn More
Mettler Toledo™ NIST/DIN pH Buffer Solution Sachets
GREENER_CHOICE
Offers the highest possible pH precision. Mettler Toledo™ NIST/DIN pH Buffer Solution Sachets are specified to three decimal places and come with a printed certificate that guarantees traceability. Individual packets guarantee fresh solution for every calibration, as well as maximum ease of use.
Brand:  Mettler Toledo™ 30111138
913.00 DKK valid until 2025-08-31
Use promo code "24157" to get your promotional price.
 View more versions of this product
Product Code. 15322275
  • 979.00 DKK / Each
Focus on Agri-Science Testing
Use promo code 24136 at checkout
View Available Promotions
Please to purchase this item. Need a web account? Register with us today!
Edge
This item is not returnable. View return policy
Description
Description
pH measurements are only as accurate as the buffer solutions used for calibration purposes. Maximum precision is guaranteed with fully traceable NIST/DIN buffers. (pH electrodes should be calibrated at least once a day prior to measurements and after every cleaning, regeneration or long-term storage.)
  • NIST/DIN pH buffers are specified and guaranteed to three decimal places, offering top precision
  • Individual sachets guarantee fresh solution for every calibration, as well as maximum ease of use
  • A test certificate and detailed label with a temperature table comes with every box
  • All SDS and labels contain information according to GHS (Globally Harmonized System)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
NIST/DIN Buffer pH 9.180 sachets 30x20mL
pH measurement, electrode calibration
Sachet
30 x 20 mL
pH Electrode
Yes
9.180 (at 25°C)
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.