Learn More
Eppendorf™ New Brunswick™ S41i CO2 Incubator Shaker
The New Brunswick S41i is a versatile, medium-sized CO2 incubator shaker ideal for growing labs with limited lab space and budget. It combines an integrated 120°C disinfection and a seamless, stainless-steel chamber, with the heavy-duty Innova triple-eccentric drive.
Brand: Eppendorf™ S41I230011
16336.06 EUR valid until 2025-12-20
Use promo code "24121" to get your promotional price.
View more versions of this product
Description
Applications
- Mammalian inoculum preparation
- Expansion of suspension mammalian cell lines, e.g. CHO, HEK293 or Vero cells for protein expression or viral vector production
- Growth of sensitive cells such mesenchymal- or induced pluripotent stem cells (MSCs or iPSCs) on microcarriers for reprogramming or exosome production
- Cultivation of adherent cells and applications that require a static and shake flask cultivation such as hybridoma cultures, the adaption of adherent to suspension cells
Features
- Two in one: Shaker platform for suspension cultures plus up to two shelves (one included) for simultaneous static CO2 incubator cultivation
- Ideal for limited lab space: Compact design fits under the bench or stacked in a small footprint
- Max. shaking speed: 25 to 400 rpm (stacked: 25 to 250/400 for top/bottom unit)
- Effective contamination protection: Integrated 120°C disinfection routine
- Easy cleaning and disinfection: Smooth, seamless chamber without a fan, easy disassembling in a few seconds, removable water tray
- High vessel flexibility and capacity: From tubes up to 5 L flasks, e.g. 10 x 1 L Erlenmeyer flasks
- Reliable process planning: Proven and tested Innova™ triple eccentric counterbalanced drive (three weight-supporting shafts) provides vibration and trouble-free operation for years
- Tight CO2 specifications for reproducible results: accuracy: ±0.2% (at 5% CO2), homogeneity: ±0.1%
- Protect your light-sensitive expression media: The double door system also helps protect the atmosphere inside and reduce CO2 consumption
- Less noise in the lab: One of the quietest CO2 incubator shakers on the market - with the Innova™ triple eccentric counterbalanced drive
Specifications
| 2.5 cm | |
| ± 1 % full scale | |
| 230 V | |
| < 8 A | |
| CO2 Orbital Shaker | |
| EU Plug | |
| ± 0.6°C at ambient 20°C to 25°C | |
| 44.0 x 69.5 x 53.5 cm (17.3 x 27.4 x 21.1 in.) | |
| 73 x 87.5 x 85 cm (28.7 x 34.4 x 33.5 in.) | |
| 192 kg | |
| 154 kg | |
| Color Touchscreen | |
| 1 Perforated Stainless Steel Shelf Installed; 1 CO2 Sensor Cover; 1 Colored protective cover; 1 Front foot shield; 2 Side foot shields; 2 Humidity Trays (stainless steel, removable for cleaning); 3 m/9.8 ft. PVC Tubing, 6 mm (1/4 in.) bore, with an inline CO2 HEPA-filter connected; 2 Hose Clips; 1 Auto-zero HEPA Filter; 1 BMS connector; 3 Rack spacers; 3 Rack feet; 2 Additional CO2 sensor covers; 1 Mains/Power Cord; 1 Operating Manual | |
| Continuous |
| 11 kg | |
| 25 to 400 rpm | |
| 4°C to 50°C | |
| 170 L | |
| CE, UL, cUL, UKCA | |
| ± 0.2°C | |
| 50/60 Hz | |
| 35.6 x 61.2 cm (14 x 24 in.) | |
| 423 lb. | |
| 340 lb. | |
| New Brunswick S41i | |
| 90 ± 5 % at 37°C | |
| 1 x USB 2.0; 1 x BMS relay contact alarm; 1 x Ethernet (only for service purposes and connecting to VisioNize Lab Suite) | |
| No |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.