Learn More
Labconco™ Neoprene Gloves for Glove Boxes
One piece, seamless construction with o-rings. Labconco™ Neoprene Gloves provide 0.015 gauge protection to the elbow.
Brand: Labconco™ 5005500
546.25 EUR valid until 2025-10-31
Use promo code "24133" to get your promotional price.
This item is not returnable.
View return policy
Description
- Neoprene Gloves
- One piece, seamless, 0.015 gauge
- With O-rings
- For use with Labconco Protector™ Glove Boxes, Precise™ Glove Boxes and XPert™ Weigh Boxes.
Specifications
| Black | |
| Powder Free | |
| Non-sterile |
| Neoprene | |
| 8.5 | |
| 1 pair |