Learn More
Cytiva Nanosep™ Centrifugal Devices with Omega™ Membrane
Use Nanosep™ devices to concentrate, purify, and desalt peptides, proteins, oligonucleotides, DNA, and RNA.
1095.00 DKK - 15750.00 DKK
Specifications
Description | EFA: 0.3 cm2 |
---|---|
Material (Membrane) | PES |
Sterility | Non-sterile |
Housing Material | Polypropylene |
Volume (Metric) | 50 to 500 μL (Sample) |
Product Code | Brand | MWCO | Includes | Color | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | MWCO | Includes | Color | Price | Quantity & Availability | |||
17164791
|
Cytiva
OD300C34 |
300 kDa | 100/Pk. | Orange |
3615.00 DKK
Pack of 100 |
Estimated Shipment: 01-08-2025
Log in to see stock. |
|||
17154791
|
Cytiva
OD300C33 |
300 kDa | 24/Pk. | Orange |
1160.00 DKK
Pack of 24 |
Estimated Shipment: 28-07-2025
Log in to see stock. |
|||
17114791
|
Cytiva
OD100C33 |
100 kDa | 24/Pk. | Clear |
1095.00 DKK
Pack of 24 |
Estimated Shipment: 28-07-2025
Log in to see stock. |
|||
17184781
|
Cytiva
OD030C34 |
30 kDa | 100/Pk. | Red |
3615.00 DKK
Pack of 100 |
Estimated Shipment: 28-07-2025
Log in to see stock. |
|||
17174781
|
Cytiva
OD030C33 |
30 kDa | 24/Pk. | Red |
1095.00 DKK
Pack of 24 |
Estimated Shipment: 28-07-2025
Log in to see stock. |
|||
17134781
|
Cytiva
OD010C33 |
10 kDa | 24/Pk. | Blue |
1095.00 DKK
Pack of 24 |
Estimated Shipment: 28-07-2025
Log in to see stock. |
|||
17104781
|
Cytiva
OD003C34 |
3 kDa | 100/Pk. | Gray |
3615.00 DKK
Pack of 100 |
Estimated Shipment: 01-08-2025
Log in to see stock. |
|||
17194771
|
Cytiva
OD003C33 |
3 kDa | 24/Pk. | Gray |
1095.00 DKK
Pack of 24 |
Estimated Shipment: 13-08-2025
Log in to see stock. |
|||
17144781
|
Cytiva
OD010C34 |
10 kDa | 100/Pk. | Blue |
3615.00 DKK
Pack of 100 |
Estimated Shipment: 29-07-2025
Log in to see stock. |
|||
17124791
|
Cytiva
OD100C34 |
100 kDa | 100/Pk. | Clear |
3615.00 DKK
Pack of 100 |
Estimated Shipment: 29-07-2025
Log in to see stock. |
|||
17154781
|
Cytiva
OD010C35 |
10 kDa | 500/Pk. | Blue |
14700.00 DKK
Pack of 500 |
Estimated Shipment: 01-08-2025
Log in to see stock. |
|||
17114781
|
Cytiva
OD003C35 |
3 kDa | 500/Pk. | Gray |
14700.00 DKK
Pack of 500 |
Estimated Shipment: 01-08-2025
Log in to see stock. |
|||
17194781
|
Cytiva
OD030C35 |
30 kDa | 500/Pk. | Red |
15750.00 DKK
Pack of 500 |
Estimated Shipment: 29-07-2025
Log in to see stock. |
|||
17134791
|
Cytiva
OD100C35 |
100 kDa | 500/Pk. | Clear |
15750.00 DKK
Pack of 500 |
Estimated Shipment: 20-08-2025
Log in to see stock. |
|||
17174791
|
Cytiva
OD300C35 |
300 kDa | 500/Pk. | Orange |
14700.00 DKK
Pack of 500 |
Estimated Shipment: 20-08-2025
Log in to see stock. |
|||
Description
- Simple, reliable,concentrating and desalting of 50 to 500 μL samples
- This product is not approved for use in medical, clinical, surgical or other patient applications
Specifications
EFA: 0.3 cm2 | |
Non-sterile | |
50 to 500 μL (Sample) |
PES | |
Polypropylene |