missing translation for 'onlineSavingsMsg'
Learn More

Implen™ NanoPhotometer™ N50-Touch Micro-Volume UV-VIS Spectrophotometer

The Implen NanoPhotometer™ N50 UV/Vis spectrophotometer for NanoVolume applications. Quantify DNA, RNA, proteins and scan other samples within seconds. Intuitive touchscreen operation, small footprint and network integration for convenient lab bench operation.

Brand:  Implen™ N50-Touch

Product Code. 15795805

  • POA / Each

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

The Implen NanoPhotometer™ N50 UV/VIS spectrophotometer is the right instrument when it comes to the quantification of nucleic acids, proteins or cell cultures as it offers scan data from 200 - 650 nm in just 0.3 - 2 μL of sample. The stand-alone design with an integrated touchscreen and small footprint make it ideal for lab bench operation.

Disturbing surface tension and evaporation effects are eliminated due to the compression of the sample. No recalibration is necessary over the entire lifetime. (Recalibration-free patented technology.)

  • Detection range dsDNA 5 ng/μL - 7,500 ng/μl
  • Detection range BSA 0.15 mg/mL - 217 mg/ml
  • Minimum sample Size 0.3 μL
  • Scan range 200 - 650 nm
  • Measure time for full scan range: 2.5 - 4.0 seconds
  • Preprogrammed applications for (labeled) nucleic acids (DNA, RNA, oligos), (labeled) proteins, protein assays (Bradford, BCA), kinetics, OD600 and manual methods
  • Auto Sample function starts automatically once sample arm is closed
  • Recalibration-free due to True Path Technology™
  • Independent from surface tension of sample due to Sample Compression Technology™
  • Sample ControlTM and Blank ControlTM for analysis of sample quality and identification of contaminants
  • Control options: Onboard with built-in 7” Touchscreen, Computer and Tablet
  • In & Output Ports 2x USB A, USB B, HDMI, Ethernet, Wi-Fi
  • Built-in file server
  • LIMS integration through REST API
  • Made in Germany
Specifications

Specifications

< 1.75% at 0.7 A at 280 nm of the reading
2x USB A, USB B, HDMI, Ethernet, WiFi
7 in. Glove Compatible Touchscreen, 1024 x 600 pixels
English, French, German, Spanish, Portuguese, Russian, Chinese, Japanese
5 nm
200 to 650 nm
Power Supply, USB Cable, Dust Cover, Software & Manual
2.5 to 4.0 sec
0.67 and 0.07 mm
0.3 to 2 μL
Windows 7, 8, 10 (32 & 64 bit); OS X, iOS; Android OS
4.5 kg
0.1 to 150 A
3648-element CCD Array
20 x 20 x 12 cm
Xenon Flash
90/250 V
Nucleic Acids, Protein UV, Protein Assays, Kinetics, OD600, Wavelength, Wavescan, Standard Curve, Absorbance Ratio, Concentration, Custom
5 to 7,500 ng/μL
1
Linux based NPOS
< 2% @ 240 nm using NaI
± 1.5 nm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
View accessory products
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only