Learn More
Implen™ NanoPhotometer™ IQ/OQ Documentation and Standard Solution
In compliance with international standards in regulated environments, documented verification is provided with Installation Qualification and Operation Qualification (IQ/OQ) Package Conforming Data to verify that instruments are working according to specifications in case of an audit.
Brand:  Implen™ N-60-Q
 View more versions of this product
Product Code. 15533214
  • 1555.00 € / Each
Please to purchase this item. Need a web account? Register with us today!
This item is not returnable. View return policy
Description
Description
  • Documentation provided on a USB flash drive as an Excel file
  • Includes NanoPhotometer™ appropriate standard solution (N-568-S or N-120-S)
Specifications
Specifications
NanoPhotometer N60
Self-calibration
NanoPhotometer
According to Internal SOP
IQ/OQ Package Conforming Data
IQ/OQ Documentation and Standard Solution (N-568-S)
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.
For Research Use Only