Learn More
Thermo Scientific™ Nalgene™ PETG Diagnostic Bottles with Closure: Sterile

Conduct sterile sampling, storage and shipment of reagents with these clear PETG bottles that are leakproof when used with provided Nalgene lined white HDPE closures.
Brand: Thermo Scientific™ 2035-0005
1613.19 DKK valid until 2025-12-20
Use promo code "24125" to get your promotional price.
Description
Conduct sterile sampling, storage and shipment of reagents and buffer solutions using Thermo Scientific™ Nalgene™ PETG Diagnostic Bottles with Closure. These bottles are constructed from non-pyrogenic and non-cytotoxic polyethylene terephthalate copolymer with lined white HDPE (high-density polyethylene) closures.
Features:- Sterile to 10-6 SAL
- Non-pyrogenic and non-cytotoxic
- 20-415 lined white high-density polyethylene closure attached to PETG vial
- Leakproof†
Ordering Information:
Serum Vials are packed in shrink-wrapped trays with closures attached.
Specifications
| 0.16 oz., 5 mL | |
| 0.16 oz. | |
| PETG | |
| 20 | |
| 20-415 mm | |
| Yes | |
| 46 mm | |
| 1.81 in. | |
| No |
| 5 mL | |
| 11 mm | |
| 100 | |
| Sterile | |
| Manufactured in compliance with ISO 13485:2016 quality requirements. Serum Vials meet the requirements of 21CFR177.1520 for food and beverage use, USP Class VI. | |
| Narrow | |
| Sample Container | |
| 22 mm |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes.
Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.