missing translation for 'onlineSavingsMsg'
Läs mer

N,N-Dimethylformamide, Reagent Grade, ≥99%, Honeywell

Produktkod. 15687790 Handla allt Honeywell Research Chemicals Produkter
Change view
Klicka för att se tillgängliga alternativ
Quantity:
500 mL
1 L
2.5 L
5 L
4 x 2.5 L
6 x 1 L
Packaging:
Poly Bottle
Förpackningsstorlek:
1 L
2.5 L
5 L
500mL
Pack of 4
Pack of 6
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Quantity Packaging unitSize
15687790 2.5 L Poly Bottle 2.5 L
15627800 500 mL Poly Bottle 500mL
15677790 1 L Poly Bottle 1 L
15637800 5 L Poly Bottle 5 L
15647800 6 x 1 L Poly Bottle Pack of 6
15617800 4 x 2.5 L Poly Bottle Pack of 4
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Denna artikel kan inte returneras. Se returpolicy
Regler: Denna produkt innehåller ett ämne som är upptaget i bilaga XVII till REACH-förordningen (EG) nr 1907/2006. Alla kunder måste fylla i en deklaration före leverans.
Produktkod. 15687790 Leverantör Honeywell Research Chemicals Leverantörsnummer D1585502.5L
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

Denna artikel kan inte returneras. Se returpolicy
Regler: Denna produkt innehåller ett ämne som är upptaget i bilaga XVII till REACH-förordningen (EG) nr 1907/2006. Alla kunder måste fylla i en deklaration före leverans.

Reagent Grade,≥99%

  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY

Kemiska identifierare

CAS 68-12-2
Molecular Formula C3H7NO
Molecular Weight (g/mol) 73.10
MDL Number MFCD00003284
InChI Key ZMXDDKWLCZADIW-UHFFFAOYSA-N
PubChem CID 6228
ChEBI CHEBI:17741
IUPAC Name N,N-dimethylformamide
SMILES CN(C)C=O

Specifikationer

Melting Point -61°C (literature)
CAS Min % ≥90.0000%
CAS Max % ≤100.0000%
Density 0.944 g/mL(lit.)
Boiling Point 152°C to 153°C
Quantity 2.5 L
Explosion Limit 0.152
Flash Point 57.5°C
Ignition Point 833°F
Packaging Poly Bottle
Linear Formula HCON(CH3)2
Vapor Pressure 2.7 mmHg (20°C)
UN Number UN2265
Beilstein 605365
Water max. 0.1%
Synonym dimethylformamide, dimethyl formamide, n,n-dimethylmethanamide, n-formyldimethylamine, formamide, n,n-dimethyl, dmf, dimethylformamid, dimetilformamide, dwumetyloformamid, dmfa
Formula Weight 73.10
Percent Purity >99%
Grade Reagent
Chemical Name or Material N,N-Dimethylformamide, Reagent Grade, ≥99%
Visa mer Visa mindre
compliance-icons compliance-icons compliance-icons
Produktbeteckning
  • N,N-Dimethylformamide
Signalord
  • Danger
Riskkategorier
  • Flammable liquid Category 3
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Reproductive toxicity Category 1B
Faroangivelser
  • H226-Flammable liquid and vapour.
  • H312 + H332-Harmful in contact with skin or if inhaled.
  • H319-Causes serious eye irritation.
  • H360D-May damage the unborn child.
Skyddsangivelse
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-¢In case of inadequate ventilation! wear respiratory protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.