missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Multi Function 3D Rotators

For continuous operation, timed operation or different combinations to ensure optimal mixing conditions for any application

Brand:  Fisherbrand™ 88861048

 View more versions of this product

Product Code. 15514080

  • ONLINE EXCLUSIVE PRICE
    850.00 €
    842.00 € / Each
    Save 8.00 €
    0.94% Off
Only null left
Estimated Shipment: 17-10-2025
Add to basket

Includes: Dimpled rubber mat and six tube-securing bands
This item is not returnable. View return policy

Description

Description

  • Rotation speed: 5 to 100 rpm for long wave motion
  • Reciprocal rotation: 0°to 360° turning angle for long and medium way motion
  • Vibration: 0° to 5° turning angle, programmable in bursts of 0 to 5 seconds, for short wave motion
  • Fits neatly into small workspaces
  • Operating temperature range of 5 to 40°C allows for use in refrigerators or non-CO2 incubators
  • Continuous or timed operation with automatic switch-off
  • Display will show last used settings, even after power has been turned off.
  • Continue to run by last set speed and the remaining time when the power recovered.
  • Audible and visual alarm: In timed mode, when the time reaches zero, alarm will sound 5 times, then the time window shows“End“ and flickers all the time. Alarm can be stopped by touch any key.

Certifications

CE

Services

2 years

TRUSTED_SUSTAINABILITY
Specifications

Specifications

23 mm
1 kg
100/240 V
2.2 lb.
EU, UK, CN Plug
266 mm
  • Orbital/360° vertical/reciprocal mode - 0 to 250 sec.
  • Vibration mode - 0- 5 sec (Vibration) with 0- 125 cycles
0 to 360°
5 to 100 rpm
5°C to 40°C
CE
306 mm
Immune precipitations, Affinity matrix , Trypsinisation, Gel straining and distaining, Antibody staining, Washes, Hybridizations, Southern blots, Western blots
2 Years
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
View accessory products
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.