Learn More
IKA™ MT 40.100 Grinding Chamber
Product Code. 15153129 Shop All IKA Products
Change view
Click to view available options
Capacity (Metric):
40 mL
Unit Size:
Pack of 100
Product Code. Capacity (Metric) unitSize
15153129 40 mL Pack of 100
1 options
Product Code. 15153129 Supplier IKA™ Supplier No. 0020001173
Only null left
Add to basket
Edge
Add to basket
The MT 40.100 disposable grinding chamber with a stainless steel beater reduces soft, medium, hard and brittle materials with a Mohs hardness of up to 5 (manganese or apatite: 5 Mohs).
The chamber is made of transparent plastic so grinding tests can be observed at any time
TRUSTED_SUSTAINABILITY
Specifications
Product Type Grinding Chamber
Capacity (Metric) 40 mL