missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ MP Magnetic Rack 24
Shop All MP Biomedicals Products
Click to view available options
:
MP Magnetic Rack 24
Description
- Magnet made of high-energy neodymium
- Small, strong and easy to handle
- For fast and efficient magnetic separations
- Design allows easy removal of rack from magnet
- Tubes can be placed on shaker or vortex during extraction process
- Good visibility of samples
- Up to 24 tubes of 1.5 or 2 mL
Storage:
- Magnet:
- Stored in a dry and cool place
- Never use > 50 °C
- Tube rack:
- Store at room temp
- Can be placed with tubes at temp as low as -20 °C for a limited time
Cleaning:
- Use with one of the following solutions:
- 70% (v/v) isopropyl alcohol
- 1% (v/v) sodium hypochlorite solution (bleach)
- 0.1 N HCl solution
Notes:
- Avoid prolonged exposure of rack to water or aqueous solutions
- Not autoclavable
Specifications
Specifications
| Capacity | 24 x 1.5 or 2 mL tubes |
| Content And Storage | Store magnet in dry, cool place, Store rack at room temp |
| For Use With (Equipment) | For use with the Magbeads FastDNA™ kit, tubes |
| Product Type | Magnetic Rack |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction