Learn More
MP Biomedicals™ Roll & Grow™ Plating Beads
Shop All MP Biomedicals ProductsDescription
Roll & Grow™ Plating Beads are used to spread 25-400μL of bacterial or yeast cells across the surface of an agar plate, providing a safer, faster alternative to the traditional method of plating with a glass rod sterilized with ethanol and flame. Sterile Roll & Grow™ Plating Beads are provided in two skirted 50mL conical tubes. Between six and seven 4mm glass beads are used per plate for a total of approximately 160 standard 100mm plates.
- Spread cells in just seconds per plate
- Eliminate hazards of flame and ethanol
- No need for fragile glass “hockey sticks”
- Consistent, even colony distribution
- Sterile and easy to use
Cell cycle; Cell proliferation; Cell metabolism; Cell viability; Cytotoxicity; Protein Trafficking; Bacteria cell culture; Cell preservation; Cell counting; Viability; Cryopreservation; Insect cell culture; Mammalian cell culture; Plant cell growth; Primary cell culture; Stem cell culture; Yeast cell culture

Specifications
Specifications
Quantity | 2 Tubes |
Material | Glass |
Product Type | Roll and Grow Plating Beads |
For Use With (Application) | Cell cycle, cell proliferation, cell metabolism, cell viability, cytotoxicity, protein trafficking, bacteria cell culture, cell preservation, cell counting, viability, cryopreservation, insect cell culture, mammalian cell culture, plant cell growth |
Includes | Skirted 50mL conical tubes |
Thickness | 4mm |
Storage Requirements | 15° to 30°C |
Sterility | Sterile |
Holds | 2 Tubes |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.