missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Accutase

Product Code. 11420705 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
100 mL
Unit Size:
100mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11420705 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11420705 Supplier MP Biomedicals™ Supplier No. 091000449

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Novel trypsin replacement that exhibits both protease and collagenolytic activities.

  • Novel trypsin replacement that exhibits both protease and collagenolytic activities for cell detachment
  • Effective in detaching primary fibroblasts, endothelial cells, neurons, tumor cell lines and insect cells
  • Maintains most cell surface antigens

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage -20°C
Product Type Cell Culture Dissociation Reagent
Sterility Sterile
pH 6.8 to 7.8
Quantity 100 mL

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.