Learn More
Miele Professional™ Mobile Injector, Module A 300
Module A 300 for the optimum loading of laboratory glassware.
Brand:  Miele Professional™ 11116900
Product Code. 15921307
  • 549.00 € / Each
Only null left
In Stock
Add to basket
This item is not returnable. View return policy
Description
Description
  • Suitable for laboratory glassware such as Erlenmeyer flasks
  • Equipped with 8 injector nozzles E 352 (6 x 220 mm)
  • Equipped with 8 locking mechanisms for nozzles E 354
  • Jet spacing 120 mm across the width, 123 mm across the depth
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Injector Module
Laboratory Washer Baskets A 100 and A 150
Two rows of four injectors
181 x 220 x 479 mm
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.