Learn More
Ohaus™ Mini Digital Vortex Mixer VXMNDG
OHAUS™ Mini Digital Vortex Mixer VXMNDG is built sturdy to provide a stable and reliable vortexing action.
Brand:  Ohaus™ 30392124
370.16 EUR valid until 2025-12-20
Use promo code "24121" to get your promotional price.
Product Code. 15702239
  • ONLINE EXCLUSIVE PRICE
    € 559.00
    € 515.00 / Each
    Save € 44.00
    7% Off
View Available Promotions
Only null left
Estimated Shipment: 28-11-2025
Add to basket
This item is not returnable. View return policy
Description
Description
  • Sturdy construction for stable and reliable vortexing action
  • Ideal for applications that demand repeatable results
  • Microprocessor control maintains set speed for strong, consistent mixing action
  • Timer will display elapsed time or, when programmed to user defined time limit, the unit will shut off when time reaches zero
  • Two modes of operation; continuous mode when using accessory attachments or touch mode which activates mixing when depressing the cup head
  • LED displays for speed and time
TRUSTED_SUSTAINABILITY
Specifications
Specifications
4.9 mm
320 V, 0.6 A, 50/60 Hz
21.00 x 12.30 cm
16.5 cm
21.00 cm
12.30 cm
500 to 2500 rpm
0.6 A
6.50 in.
8.30 in.
4.80 in.
VXMNDG
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.