missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ohaus™ Mini Analog Vortex Mixer VXMNAL
OHAUS™ Mini Analog Vortex Mixer VXMNAL is built sturdy to provide a stable and reliable vortexing action.
Brand: Ohaus™ 30392117
1801.66 DKK valid until 2025-12-20
Use promo code "24121" to get your promotional price.
This item is not returnable.
View return policy
Description
- Sturdy design and construction to provide stable and reliable vortexing action
- Control allows low rpm startup for gentle shaking or high speed mixing for vigorous vortexing of samples
- Two modes of operation; 'Continuous' mode when using accessory attachments, or 'Touch' mode which activates mixing when depressing the cup head
- Sturdy design
- Variable, analogue speed control

Specifications
4.9 mm | |
230 V, 50/60 Hz, 0.6 A | |
Mini Analog Vortex Mixer | |
21.00 x 12.30 cm | |
16.5 cm | |
21.00 cm | |
12.30 cm |
300 to 2500 rpm | |
0.6 A | |
EU Plug | |
6.50 in. | |
8.30 in. | |
4.80 in. | |
VXMNAL, EU |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction