missing translation for 'onlineSavingsMsg'
Learn More

Micronova™ Irradiated IPA Resistant LabMarker
BIOPHARMA

Product Code. 12857053 Shop All Micronova Products
Click to view available options
Color:
Black
Unit Size:
Pack of 10

Product Code. 12857053

Brand: Micronova™ undefined

Only null left
Add to basket
Edge
Add to basket

A fine tipped permanent marker with alcohol-resistant ink that marks on cold or wet surfaces, plastic bags and disposable lab-ware.

With permanent, quick drying black ink that is resistant to IPA

The Micronova Irradiated IPA Resistant LabMarker features permanent, quick drying, alcohol-resistant ink that marks on a range of surfaces and will not smear on reusable glass, metal or porcelain, but can be scrubbed off. Available gamma irradiated for aseptic areas.

  • Fine-tip, black permanent marker
  • Quick drying, alcohol-resistant ink
  • Marks on cold or wet surfaces, plastic bags, and disposable lab-ware
  • Smear-resistant on glass, metal and porcelain, but can be scrubbed off
  • Available gamma irradiated for aseptic areas

Specifications

Material HDPE
Color Black
Length (English) 5.25 in.
Autoclavable Not Autoclavable
Disposable Reusable
For Use With (Application) Cleanroom and Aseptic Environment Documentation and Labeling
Packaging 10/Pk.
Product Type Cleanroom Pen
Quantity Ea.
Sterility Sterile
Tip Style Fine
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.