missing translation for 'onlineSavingsMsg'
Learn More

Micronova™ IPA Resistant LabMarker
BIOPHARMA

Product Code. 12877053 Shop All Micronova Products
Change view
Click to view available options
Color:
Black
Unit Size:
Pack of 10
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color unitSize
12877053 Black Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 12877053 Supplier Micronova™ Supplier No. PEN40
Only null left
Add to basket
Edge
Add to basket

A fine tipped permanent marker with alcohol-resistant ink that marks on cold or wet surfaces, plastic bags and disposable lab-ware.

With permanent, quick drying black ink that is resistant to IPA

The Micronova IPA Resistant LabMarker features permanent, quick drying, alcohol-resistant ink that marks on a range of surfaces and will not smear on reusable glass, metal or porcelain, but can be scrubbed off. Available gamma irradiated for aseptic areas.

  • Fine-tip, black permanent marker
  • Quick drying, alcohol-resistant ink
  • Marks on cold or wet surfaces, plastic bags, and disposable lab-ware
  • Smear-resistant on glass, metal and porcelain, but can be scrubbed off
  • Available gamma irradiated for aseptic areas

Specifications

Tip Style Fine
Product Type Cleanroom Pen
Color Black
Autoclavable Not Autoclavable
Sterility Non-sterile
Material (Barrel) High-Density Polypropylene
Disposable Reusable
For Use With (Application) Cleanroom and Aseptic Environment Documentation and Labeling
No. per Pack 10
Quantity Ea.
Capacity (Metric) 2 mL
Length (English) 5.25 in.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.