missing translation for 'onlineSavingsMsg'
Learn More
Learn More
METTLER TOLEDO™ InLab™ Dairy pH Electrode
A pH electrode featuring a triple ceramic junction, designed for protein containing samples such as dairy products, milk and cream.
Brand: METTLER TOLEDO™ 59904591
This item is not returnable.
View return policy
Description
- For use with METTLER TOLEDO SevenExcellence and SevenCompact meters as well as other brands. Connection cables may be required.
- Capture electrode calibration and results with LabX software, designed for advanced data management and 21CFR part 11 regulatory compliance.
- Compatible with InMotion Autosampler to automate measurement and cleaning of up to 303 samples.
Featured Applications
- Milk and cream
- Protein containing samples
- Emulsion
Accessories & Care
- Triple ceramic junction
- A41 membrane glass
- Refillable FRISCOLYT-B™ (51343185): Keep probe filled with electrolyte for accurate results
- Store in 3M KCl (51350072) or InLab Storage Solution (30111142)
- Recommended for use with METTLER TOLEDO buffers (Sachets: 51302080, Bottles: 63056116)

Specifications
pH electrode InLab Dairy | |
Triple/Ceramic | |
0 to 12 | |
Combination Electrode | |
Used to puncture soft media or in aqueous samples | |
ARGENTHAL | |
InLab™ Dairy | |
0°F to 212°F | |
4.7 in. | |
0.47 in. |
S7 | |
Glass | |
0°C to 100°C | |
FRISCOLYT-B | |
S7 to BNC (30281915) | |
A41 Glass | |
3mol/L KCl | |
0°C to 100°C | |
120 mm | |
12 mm |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction