missing translation for 'onlineSavingsMsg'
Learn More

Mettler Toledo™ Balance Printer Accessories: Printer Paper

Product Code. 10095511 Shop All Mettler Toledo Products
Change view
Click to view available options
Quantity:
5 Rolls/Pk.
Unit Size:
Pack of 5
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10095511 5 Rolls/Pk. Pack of 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10095511 Supplier Mettler Toledo™ Supplier No. 72456

Please to purchase this item. Need a web account? Register with us today!

Printer paper for use with Mettler Toledo™ balances.

  • Color: White
  • Five rolls per pack
  • For use with Mettler Toledo balances: LC-P45, LC-P43, HA-P43, HR-73P
TRUSTED_SUSTAINABILITY

Specifications

For Use With (Equipment) LC-P45, LC-P43, HA-P43, HR-73P Printer
Quantity 5 Rolls/Pk.
Type Printer Paper
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.