missing translation for 'onlineSavingsMsg'
Learn More

Methyl sulfoxide-d6, for NMR, packaged in 0.75 ml ampoules, 100.0 atom % D

Product Code. 10791541 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
7.5 mL
Unit Size:
7.50mL
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

CAS: 2206-27-1 | C2H6OS | 84.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For NMR

Specifications

Physical Form Liquid
Color Colorless
Density 1.1900g/mL
Deuterium (D) 99.95% min.
Flash Point 88°C
Specific Gravity 1.19
Viscosity 2.4 mPa.s (20°C)
Water 0.02% max. (Coulometric)
Quantity 7.5 mL
CAS 2206-27-1
Melting Point 20.2°C
Boiling Point 190.0°C
Molecular Formula C2H6OS
Linear Formula (CD3)2SO
MDL Number MFCD00002090
Beilstein 01, IV, 1279
Fieser 13,124; 16,177
Synonym dimethyl sulfoxide-d6, dmso-d6, dimethylsulfoxide-d6, deuterated dmso, methane-d3,sulfinylbis-9ci, methane-d3, sulfinylbis, hexadeuterodimethyl sulfoxide, methane-d3-, sulfinylbis, methanesulfinylmethyl hydrogen, dimethyl-d6 sulfoxide
InChI Key IAZDPXIOMUYVGZ-WFGJKAKNSA-N
SMILES [2H]C([2H])([2H])S(=O)C([2H])([2H])[2H]
IUPAC Name trideuterio(trideuteriomethylsulfinyl)methane
Molecular Weight (g/mol) 84.17
PubChem CID 75151
Formula Weight 84.18
Grade NMR
Chemical Name or Material Methyl sulfoxide-d6, 100.0 atom % D
Show More Show Less
Product Identifier
  • Methyl sulfoxide-d6

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.