missing translation for 'onlineSavingsMsg'
Learn More

Methanol-d4, for NMR, with 0.03% TMS, in 0.75 ml ampoules, 99.8 atom % D

Product Code. 10525671 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
7.5 mL
Unit Size:
7.50mL
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

CAS: 811-98-3 | CH4O | 36.066 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For NMR

Specifications

Physical Form Liquid
Color Colorless
Density 0.8880g/mL
Deuterium (D) 99.75% min.
Flash Point 12°C
Refractive Index 1.3262
Water 0.05% max. (Coulometric)
Quantity 7.5 mL
CAS 811-98-3
Melting Point -99.0°C
Boiling Point 65.0°C
Molecular Formula CH4O
Linear Formula CD3OD
MDL Number MFCD00044637
Beilstein 01, III, 1187
Synonym methanol-d4, perdeuteromethanol, tetradeuteromethanol, methan-d3-ol-d, 2h4 methanol, methyl alcohol-d4, cd3od, methyl-d3 alcohol-d, trideuterio deuteriooxy methane, methanol-d4, 99.8 atom % d
InChI Key OKKJLVBELUTLKV-MZCSYVLQSA-N
SMILES CO
IUPAC Name trideuterio(deuteriooxy)methane
Molecular Weight (g/mol) 36.066
PubChem CID 71568
Formula Weight 36.06
Grade NMR
Chemical Name or Material Methanol-d4, 99.8 atom% D
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Methanol-d4
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Acute toxicity Category 3
  • Specific target organ toxicity after single exposure Category 1
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H370-Causes damage to organs.
  • H301-Toxic if swallowed.
  • H311-Toxic in contact with skin.
  • H331-Toxic if inhaled.
Precautionary Statement
  • P310-Immediately call a POISON CENTER/doctor/.
  • P308+P311-IF exposed or concerned: Call a POISON CENTER/doctor/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.